Learn More
BPS Bioscience Inc Firefly Luciferase Lentivirus (G418, Hygromycin and Puromycin)
SDP
Catalog No. NC1839850
Encompass
The Firefly Luciferase Lentivirus are replication incompetent, HIV-based, VSV-G pseudotyped lentiviral particles that are ready to be transduced into almost all types mammalian cells, including primary and non-dividing cells. This virus constitutively express firefly luciferase under a CMV promoter.
Catalog No. NC1839850 Supplier BPS Bioscience Inc Supplier No. 79692H
The Firefly Luciferase Lentivirus are replication incompetent, HIV-based, VSV-G pseudotyped lentiviral particles that are ready to be transduced into almost all types mammalian cells, including primary and non-dividing cells. This virus constitutively express firefly luciferase under a CMV promoter.