Learn More
Apexbio Technology LLC EZ Cap™ Firefly Luciferase mRNA, 100ug
SDP
Catalog No. 501998497
Encompass_Preferred
EZ Cap™ Firefly Luciferase mRNA will express luciferase protein once entering cells, which is initially extracted from firefly Photinus pyralis. This enzyme catalyzes ATP-dependent D-luciferin oxidation and lead to yield chemiluminescence at about 560 nm wavelength. Firefly Luciferase is a frequently used bioluminescent reporter for gene regulation and function study. It is applicable in assays for mRNA delivery, translation efficiency, cell viability and in vivo imaging etc.EZ Cap™ Firefly Luciferase mRNA is provided at a concentration of 1mg/ml. It is co-transcriptional capped by EZ Cap™ Reagent AG (3' OMe) (Catalog No.B8178) which generates a cap 1 structure with high efficiency. Cap 1 structure is more ideal for mammalian systems and possesses higher transcription efficiency than Cap 0 structure (ARCA and mCap). Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8497 Supplier Apexbio Technology LLC Supplier No. R10180.1
EZ Cap™ Firefly Luciferase mRNA will express luciferase protein once entering cells, which is initially extracted from firefly Photinus pyralis. This enzyme catalyzes ATP-dependent D-luciferin oxidation and lead to yield chemiluminescence at about 560 nm wavelength. Firefly Luciferase is a frequently used bioluminescent reporter for gene regulation and function study. It is applicable in assays for mRNA delivery, translation efficiency, cell viability and in vivo imaging etc.EZ Cap™ Firefly Luciferase mRNA is provided at a concentration of 1mg/ml. It is co-transcriptional capped by EZ Cap™ Reagent AG (3' OMe) (Catalog No.B8178) which generates a cap 1 structure with high efficiency. Cap 1 structure is more ideal for mammalian systems and possesses higher transcription efficiency than Cap 0 structure (ARCA and mCap). Other sizes are also available. Please inqury us for quote.