Learn More
Apexbio Technology LLC EZ Cap™ Firefly Luciferase mRNA (5-moUTP), 5x1mg (1mg/mL)
SDP
Catalog No. 501998324
Encompass_Preferred
EZ Cap™ Firefly Luciferase mRNA (5-moUTP) will express luciferase protein once entering cells, which is initially extracted from firefly Photinus pyralis. This enzyme catalyzes ATP-dependent D-luciferin oxidation and lead to yield chemiluminescence at about 560 nm wavelength. Firefly Luciferase is a frequently used bioluminescent reporter for gene regulation and function study. It is applicable in assays for mRNA delivery, translation efficiency, cell viability and in vivo imaging etc. EZ Cap™ Firefly Luciferase mRNA (5-moUTP) is provided at a concentration of 1mg/ml. It is co-transcriptional capped by EZ Cap™ Reagent AG (3' OMe) (Catalog No.B8178) which generates a cap 1 structure with high efficiency. Other sizes are also available. Please inqury us for quote.
Catalog No. 50-199-8324 Supplier Apexbio Technology LLC Supplier No. R10135
EZ Cap™ Firefly Luciferase mRNA (5-moUTP) will express luciferase protein once entering cells, which is initially extracted from firefly Photinus pyralis. This enzyme catalyzes ATP-dependent D-luciferin oxidation and lead to yield chemiluminescence at about 560 nm wavelength. Firefly Luciferase is a frequently used bioluminescent reporter for gene regulation and function study. It is applicable in assays for mRNA delivery, translation efficiency, cell viability and in vivo imaging etc. EZ Cap™ Firefly Luciferase mRNA (5-moUTP) is provided at a concentration of 1mg/ml. It is co-transcriptional capped by EZ Cap™ Reagent AG (3' OMe) (Catalog No.B8178) which generates a cap 1 structure with high efficiency. Other sizes are also available. Please inqury us for quote.