Learn More
Cayman Chemical Firefly Luciferase mRNA Cap-
SDP
Catalog No. 502941984
Encompass_Preferred
An mRNA for the expression of luciferase; encapsulation in LNPs can be used for mRNA delivery and expression of luciferase protein in vitro or in vivo
Catalog No. 50-294-1984 Supplier Cayman Chemical Supplier No. 39801100 UG
An mRNA for the expression of luciferase; encapsulation in LNPs can be used for mRNA delivery and expression of luciferase protein in vitro or in vivo