Learn More
Medchemexpress LLC Firefly luciferase mRNA-LNP | 500 UG
SDP
Catalog No. 5000373513
Encompass_Preferred
Firefly luciferase mRNA-LNP is a lipid nanoparticle (LNP) designed to protect and transport firefly luciferase mRNA to cells. As a bioluminescent reporter gene, it expresses firefly luciferase protein for gene regulation and functional studies, finding application in RNA delivery, cell viability, and translation efficiency research.
  • LNP provides protection and transport of mRNA to cells.
  • Expresses firefly luciferase protein for gene regulation and functional studies.
  • Useful for promoter activity detection and dual fluorescent molecular complementation.
  • Demonstrates increased transfection efficiency in HepG2 cells.
  • Enables luminescent flux and protein production in various mouse models (e.g., C57BL/6, BALB/c).
  • Intravenous delivery offers peak protein production; intramuscular and intradermal deliveries ensure longer duration of translation.
Catalog No. 50-003-73513 Supplier Medchemexpress LLC Supplier No. HY153229500UG
Firefly luciferase mRNA-LNP is a lipid nanoparticle (LNP) designed to protect and transport firefly luciferase mRNA to cells. As a bioluminescent reporter gene, it expresses firefly luciferase protein for gene regulation and functional studies, finding application in RNA delivery, cell viability, and translation efficiency research.
  • LNP provides protection and transport of mRNA to cells.
  • Expresses firefly luciferase protein for gene regulation and functional studies.
  • Useful for promoter activity detection and dual fluorescent molecular complementation.
  • Demonstrates increased transfection efficiency in HepG2 cells.
  • Enables luminescent flux and protein production in various mouse models (e.g., C57BL/6, BALB/c).
  • Intravenous delivery offers peak protein production; intramuscular and intradermal deliveries ensure longer duration of translation.