Learn More
Cayman Chemical Firefly Luciferase mRNA (Cap-1; mo5U), 1 mg
SDP
Catalog No. 502942123
Encompass_Preferred
An mRNA for the expression of luciferase; encapsulation in LNPs can be used for mRNA delivery and expression of luciferase protein in vitro or in vivo
Catalog No. 50-294-2123 Supplier Cayman Chemical Supplier No. 398011 MG
An mRNA for the expression of luciferase; encapsulation in LNPs can be used for mRNA delivery and expression of luciferase protein in vitro or in vivo