Learn More
Mayflower Bioscience FIREPol DNA Polymerase
SDP
Catalog No. 502963911
Encompass
FIREPol is a highly processive, thermostable DNA Polymerase. Due to its genetic modifications, it has enhanced stability at room temperature with no activity loss for up to 1 month. Recommended for routine applications (fragment up to 3 kb from genomic DNA). Possesses 5′→3′ polymerase and 5′→3′ exonuclease activity, as well as a non-template-dependent terminal transferase activity, but lacks a 3′→5′ exonuclease (proofreading) activity making the generated product suitable for TA-cloning. The fidelity of FIREPol is similar to a regular Taq DNA Polymerase (error rate per nucleotide ca 2.5 x10-5).
Catalog No. 50-296-3911 Supplier Mayflower Bioscience Supplier No. 0101KIT00500
FIREPol is a highly processive, thermostable DNA Polymerase. Due to its genetic modifications, it has enhanced stability at room temperature with no activity loss for up to 1 month. Recommended for routine applications (fragment up to 3 kb from genomic DNA). Possesses 5′→3′ polymerase and 5′→3′ exonuclease activity, as well as a non-template-dependent terminal transferase activity, but lacks a 3′→5′ exonuclease (proofreading) activity making the generated product suitable for TA-cloning. The fidelity of FIREPol is similar to a regular Taq DNA Polymerase (error rate per nucleotide ca 2.5 x10-5).