Learn More
Bioworld Firepol (Thermostable TAQ), 4000 Unit(s)
Catalog No. 50197022
Encompass_Preferred
A thermostable DNA polymerase FirePol is similar to Taq DNA polymerase I (homology 98%). 3' to 5' exonuclease activity. Has the "extendase activity", allowing TA cloning. Due to the unique modifications and purification technology it is very thermostable at high temperatures. Can be used in all applications involving polymerase chain reaction (PCR). Contains necessary reaction buffers and MgCl2.
Catalog No. 50-197-022 Supplier Bioworld Supplier No. 111400052
A thermostable DNA polymerase FirePol is similar to Taq DNA polymerase I (homology 98%). 3' to 5' exonuclease activity. Has the "extendase activity", allowing TA cloning. Due to the unique modifications and purification technology it is very thermostable at high temperatures. Can be used in all applications involving polymerase chain reaction (PCR). Contains necessary reaction buffers and MgCl2.