Learn More
Mayflower Bioscience FIREScript RTcDNA synthesisKIT
SDP
Catalog No. 502963913
Encompass
FIREScript Reverse Transcriptase (RT) is a genetically engineered MMLV (Moloney Murine Leukemia Virus) based Reverse Transcriptase. This is an RNA‐directed DNA polymerase that can synthesize a complementary DNA strand from ssRNA or ssDNA and is active over a broad range of reaction temperatures from 37°C to 60°C. FIREScript RT is a robust enzyme for RNA detection and has enhanced stability at room temperature with no activity loss for up to 1 month. This RT contains a functional RNase H domain which can increase the sensitivity of RT‐qPCR (quantitative reverse transcription PCR).
Catalog No. 50-296-3913 Supplier Mayflower Bioscience Supplier No. 061500050
FIREScript Reverse Transcriptase (RT) is a genetically engineered MMLV (Moloney Murine Leukemia Virus) based Reverse Transcriptase. This is an RNA‐directed DNA polymerase that can synthesize a complementary DNA strand from ssRNA or ssDNA and is active over a broad range of reaction temperatures from 37°C to 60°C. FIREScript RT is a robust enzyme for RNA detection and has enhanced stability at room temperature with no activity loss for up to 1 month. This RT contains a functional RNase H domain which can increase the sensitivity of RT‐qPCR (quantitative reverse transcription PCR).