Learn More
Ace Glass, Inc. Firestone valve, #15 O-ring joint connection to system
SDP
Catalog No. 50979872
Encompass_Preferred
Firestone rapid purge valve with #15 o-ring joint connection for 100% replacement of air in reaction vessels with any desired gas. No mercury used, no reducing valve needed on gas source, and no high vacuum source necessary. Eliminates need to watch manometers, hand control gas flow, or install warning systems to prevent accidents. Expensive gases are conserved because once purging is complete, the flow can be cut almost to zero.
Catalog No. 50-979-872 Supplier Ace Glass, Inc. Supplier No. 876630
Firestone rapid purge valve with #15 o-ring joint connection for 100% replacement of air in reaction vessels with any desired gas. No mercury used, no reducing valve needed on gas source, and no high vacuum source necessary. Eliminates need to watch manometers, hand control gas flow, or install warning systems to prevent accidents. Expensive gases are conserved because once purging is complete, the flow can be cut almost to zero.