Learn More
Ace Glass, Inc. Firestone valve, hose connection to system
SDP
Catalog No. 509558584
Encompass_Preferred
Firestone rapid purge valve with hose connection for 100% replacement of air in reaction vessels with any desired gas. No mercury used, no reducing valve needed on gas source, and no high vacuum source necessary. Eliminates need to watch manometers, hand control gas flow, or install warning systems to prevent accidents. Expensive gases are conserved because once purging is complete, the flow can be cut almost to zero.
Catalog No. 50-955-8584 Supplier Ace Glass, Inc. Supplier No. 876612
Firestone rapid purge valve with hose connection for 100% replacement of air in reaction vessels with any desired gas. No mercury used, no reducing valve needed on gas source, and no high vacuum source necessary. Eliminates need to watch manometers, hand control gas flow, or install warning systems to prevent accidents. Expensive gases are conserved because once purging is complete, the flow can be cut almost to zero.