Learn More
Shelby™ Firewall Steamblock™ Gloves
Catalog No. 19821497 Shop All Shelby Specialty Gloves Products
Click to view available options
Size:
Small
Large
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
19-821-497 Large
19-821-495 Small
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-821-497 Supplier Shelby™ Supplier No. 5229L
Shipboard firefighting gloves protect the back of the hand from steam
  • Koala-tanned, leather wristlet-style gloves incorporate an outer shell, moisture barrier (liquid/fluid) and thermal liner
  • Includes a Steamblock™ insulating pad between the outer shell and moisture barrier on the back of the hand
  • Glove extends circumferentially not less than 2 in. (5 cm) beyond the wrist crease
  • Close-fitting design restricts the entry of embers and other debris
Compliance
NFPA 1971-2013
Specifications
Certifications/Compliance NFPA 1971, CE PPE, EN 659:1996, EN 388:1994
Material Cowhide Leather
Product Type Structural Firefighting Gloves
Size Large
Glove Cuff Style Knit
Liner Material Thermal