Learn More
Carbosynth LLC. H-Phe-Ile-Arg-Gly-Gly-Met-Tyr-Glu-Gly-Lys-Lys-OH, 5mg, MW: 1285.54, Molecular formula: C58H92N16O15S
SDP
Supplier:  Carbosynth LLC. TSP3874PI5MG
Encompass_Preferred
FIRGGMYEGKK, Application: Control Peptide for CD47-Binding Peptide 7N3 (TSP-3873-PI) , Salt Form:, Trifluoroacetate Form, Storage Temperature: -20 °C, References: S. Kaur, et al., J. Biol. Chem., 285, 38923 (2010).
Catalog No. 50-168-7208