Learn More
Grainger HONEYWELL HOWARD LEIGHT, Ear Plugs Cylinder, 22PN99
Catalog No. NC2825067
Encompass_Preferred
The Honeywell Howard Leight Ear Plugs Cylinder contains 200 pairs of Max Lite earplugs. The earplugs have a contoured T-shape that fits easily in the ear canal. They are made of polyurethane foam and have a Noise Reduction Rating (NRR) of 33 decibels. The earplugs are designed to block hazardous noise levels to help prevent noise-induced hearing loss. They come pre-formed in a compact cylinder dispenser for convenient access and portability. The earplugs are disposable and intended for single use application.
Catalog No. NC2825067 Supplier Grainger Supplier No. 22PN99
The Honeywell Howard Leight Ear Plugs Cylinder contains 200 pairs of Max Lite earplugs. The earplugs have a contoured T-shape that fits easily in the ear canal. They are made of polyurethane foam and have a Noise Reduction Rating (NRR) of 33 decibels. The earplugs are designed to block hazardous noise levels to help prevent noise-induced hearing loss. They come pre-formed in a compact cylinder dispenser for convenient access and portability. The earplugs are disposable and intended for single use application.