Learn More
Medchemexpress LLC Firmonertinib (mesylate) | 2130958-55-1 | 100.0% | 664.70 | 100 MG
SDP
Catalog No. 5000438764
Encompass_Preferred
Firmonertinib mesylate is an orally active, mutant-selective, and blood-brain barrier penetrant EGFR inhibitor. It inhibits EGFR active mutations as well as the T790M acquired resistant mutation. Firmonertinib mesylate has the potential for the research of cancer diseases, especially advanced non-small cell lung cancer (NSCLC) with EGFR ex20ins mutation.
  • Orally active
  • Mutant-selective
  • Blood-brain barrier penetrant EGFR inhibitor
  • Inhibits EGFR active mutations
  • Inhibits T790M acquired resistant mutation
  • Potential for research of cancer diseases, especially advanced non-small cell lung cancer (NSCLC) with EGFR ex20ins mutation
Catalog No. 50-004-38764 Supplier Medchemexpress LLC Supplier No. HY112870A100MG
Firmonertinib mesylate is an orally active, mutant-selective, and blood-brain barrier penetrant EGFR inhibitor. It inhibits EGFR active mutations as well as the T790M acquired resistant mutation. Firmonertinib mesylate has the potential for the research of cancer diseases, especially advanced non-small cell lung cancer (NSCLC) with EGFR ex20ins mutation.
  • Orally active
  • Mutant-selective
  • Blood-brain barrier penetrant EGFR inhibitor
  • Inhibits EGFR active mutations
  • Inhibits T790M acquired resistant mutation
  • Potential for research of cancer diseases, especially advanced non-small cell lung cancer (NSCLC) with EGFR ex20ins mutation