Learn More
Medchemexpress LLC Firmonertinib mesylate | 2130958-55-1 | 99.9% | 664.70 | 25 MG
SDP
Catalog No. 5000439316
Encompass_Preferred
Firmonertinib mesylate is an orally active, mutant-selective, and blood-brain barrier penetrant EGFR inhibitor. It inhibits EGFR active mutations and the T790M acquired resistant mutation. This compound has potential for research into cancer diseases, particularly advanced non-small cell lung cancer (NSCLC) with EGFR ex20ins mutation.
  • Orally active
  • Mutant-selective
  • Blood-brain barrier penetrant EGFR inhibitor
  • Inhibits EGFR active mutations
  • Inhibits T790M acquired resistant mutation
  • Potential for research of cancer diseases, especially advanced non-small cell lung cancer (NSCLC) with EGFR ex20ins mutation
Catalog No. 50-004-39316 Supplier Medchemexpress LLC Supplier No. 25MGHY112870A
Firmonertinib mesylate is an orally active, mutant-selective, and blood-brain barrier penetrant EGFR inhibitor. It inhibits EGFR active mutations and the T790M acquired resistant mutation. This compound has potential for research into cancer diseases, particularly advanced non-small cell lung cancer (NSCLC) with EGFR ex20ins mutation.
  • Orally active
  • Mutant-selective
  • Blood-brain barrier penetrant EGFR inhibitor
  • Inhibits EGFR active mutations
  • Inhibits T790M acquired resistant mutation
  • Potential for research of cancer diseases, especially advanced non-small cell lung cancer (NSCLC) with EGFR ex20ins mutation