Learn More
Medchemexpress LLC HY-16901 10mg Medchemexpress, Firsocostat CAS:1434635-54-7 Purity:>98%
SDP
Catalog No. 502029218
Encompass_Preferred
Medchemexpress, HY-16901 10mg Firsocostat CAS:1434635-54-7 Firsocostat (ND-630; GS-0976; NDI-010976) is an acetyl-CoA carboxylase (ACC) inhibitor; inhibits human ACC1 and ACC2 with IC50 values of 2.1 and 6.1 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9218 Supplier Medchemexpress LLC Supplier No. HY1690110MG
Medchemexpress, HY-16901 10mg Firsocostat CAS:1434635-54-7 Firsocostat (ND-630; GS-0976; NDI-010976) is an acetyl-CoA carboxylase (ACC) inhibitor; inhibits human ACC1 and ACC2 with IC50 values of 2.1 and 6.1 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.