Learn More
Medchemexpress LLC HY-16901A 5mg Medchemexpress, Firsocostat S enantiomer CAS: Purity:>98%
SDP
Catalog No. 501969977
Encompass_Preferred
Medchemexpress, HY-16901A 5mg Firsocostat S enantiomer CAS: Firsocostat S enantiomer (ND-630 S enantiomer) is the less active S enantiomer of Firsocostat. Firsocostat is an acetyl-CoA carboxylase (ACC) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9977 Supplier Medchemexpress LLC Supplier No. HY16901A5MG
Medchemexpress, HY-16901A 5mg Firsocostat S enantiomer CAS: Firsocostat S enantiomer (ND-630 S enantiomer) is the less active S enantiomer of Firsocostat. Firsocostat is an acetyl-CoA carboxylase (ACC) inhibitor. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.