Learn More
Med Vet International McKesson 50-Person First Aid Kit, 30325, Each
Supplier:  Med Vet International 1066512EA
Encompass_Preferred
First Aid Kit McKesson 50 Person Plastic Case Bandaging options include: 60 1 x 3 inch plastic bandages; 10 fingertip fabric bandages; 10 knuckle fabric bandages; 1/2 inch x 5 yard first aid tape Gauze items include: two 2-inch conforming gauze rolls; six 3 x 3 inch gauze dressing; and 4-inch conforming gauze roll Wound cleansing items include: 20 alcohol wipes; 12 triple antibiotic ointment packets; 15 BZK (benzalkonium chloride) antiseptic towelettes; four povidone-iodine infection control wipes; 12 0.9-gram first aid/burn cream packets Compression items include: 2 inch x 5 yard elastic bandage; 4 x 5 inch cold compress, 40 x 40 x 56 inch triangular sling/bandage, two 5 x 9 inch trauma pads Eye injury items include: two sterile eye pads and 1-ounce eyewash Tools include CPR mask with one-way valve; two pair nitrile gloves; scissors and tweezers Remaining items are comprised of 4 x 4 inch burn dressing, three insect count sting relief wipes and 10 0.9-gram hand sanitizer packets
Catalog No. 50-304-3700