Learn More
Enterprise Technology Solutions First Aid Only, Inc Alcohol Cleansing Pads, 20/BX, White
SDP
Catalog No. 5000828130
Encompass_Preferred
Prevent infections with these single-use, alcohol cleansing pads. Each pad is saturated with 70 percent isopropyl alcohol to kill germs and infections surrounding a wound. Pads come individually wrapped in a convenient dispenser box designed to fit securely into place in all of the First Aid Only Workplace and Foodservice Cabinets. Box also includes a SmartTab ezRefill indicator tab that alerts you when it's time to reorder. This item meets the ANSI/ISEA Z308.1-2009 standard.
Catalog No. 50-008-28130 Supplier Enterprise Technology Solutions Supplier No. FAOFAE4001
Prevent infections with these single-use, alcohol cleansing pads. Each pad is saturated with 70 percent isopropyl alcohol to kill germs and infections surrounding a wound. Pads come individually wrapped in a convenient dispenser box designed to fit securely into place in all of the First Aid Only Workplace and Foodservice Cabinets. Box also includes a SmartTab ezRefill indicator tab that alerts you when it's time to reorder. This item meets the ANSI/ISEA Z308.1-2009 standard.