Learn More
Enterprise Technology Solutions First Aid Only, Inc BBP Unitized Spill Clean Up Kit, White
SDP
Catalog No. 5000827805
Encompass_Preferred
Blood-borne pathogen (BBP)/personal protection kit with easy-to-open boxes features two emergency response kits in one to protect both you and the injured person. Personal protection pack includes a CPR face mask, gloves, isolation gown, eye and face shield, biohazard bag and BZK clean up wipes to safely perform life-saving CPR. Cleanup pack for body fluid spills contains an assortment of disposable tools and personal protective supplies that are suitable for the cleanup of blood-borne pathogens or other potentially infectious substances. Cleanup pack includes paper towels, chloride antiseptic towelettes, disinfectant cloth, scoop, spatula, nitrile exam gloves, absorbent powder, biohazard bag, unmarked bag and twist ties.
Catalog No. 50-008-27805 Supplier Enterprise Technology Solutions Supplier No. FAO3065
Blood-borne pathogen (BBP)/personal protection kit with easy-to-open boxes features two emergency response kits in one to protect both you and the injured person. Personal protection pack includes a CPR face mask, gloves, isolation gown, eye and face shield, biohazard bag and BZK clean up wipes to safely perform life-saving CPR. Cleanup pack for body fluid spills contains an assortment of disposable tools and personal protective supplies that are suitable for the cleanup of blood-borne pathogens or other potentially infectious substances. Cleanup pack includes paper towels, chloride antiseptic towelettes, disinfectant cloth, scoop, spatula, nitrile exam gloves, absorbent powder, biohazard bag, unmarked bag and twist ties.