Learn More
Enterprise Technology Solutions First Aid Only, Inc Eyewash Additive,Concentrate,8 oz,CL
SDP
Catalog No. 5000828328
Encompass_Preferred
Eyewash Additive Concentrate creates 5 to 20 gallons of eyewash per bottle. It's perfect for use with eyewash stations. Breakaway seal lets you know if the preservative was tampered with or previously opened.
Catalog No. 50-008-28328 Supplier Enterprise Technology Solutions Supplier No. FAO90496
Eyewash Additive Concentrate creates 5 to 20 gallons of eyewash per bottle. It's perfect for use with eyewash stations. Breakaway seal lets you know if the preservative was tampered with or previously opened.