Learn More
Enterprise Technology Solutions First Aid Only, Inc Eyewash Station,Self-contained,16 Gallon,CL
SDP
Catalog No. 5000828326
Encompass_Preferred
Physicians Care Self-Contained Eyewash Station quickly and effectively cleans debris from your eyes. One-step solution delivers up to 15 minutes of non-injurious flow of eyewash solution. Eyewash activates in 1 second and complies with ANSI Z358.1 standard for self-contained eyewash stations.
Catalog No. 50-008-28326 Supplier Enterprise Technology Solutions Supplier No. FAO91288
Physicians Care Self-Contained Eyewash Station quickly and effectively cleans debris from your eyes. One-step solution delivers up to 15 minutes of non-injurious flow of eyewash solution. Eyewash activates in 1 second and complies with ANSI Z358.1 standard for self-contained eyewash stations.