Learn More
Enterprise Technology Solutions First Aid Only, Inc Trauma Pad Refill, 5"x9", White
SDP
Catalog No. 5000827883
Encompass_Preferred
All-purpose trauma or combine dressing pad is ideal to stop bleeding associated with deep lacerations, abrasions, burns, penetration wounds and fractures. Sterile pad is individually wrapped to prevent contamination. Color-coded, unitized refill is designed to fit neatly into unitized first aid stations or kits. It also meets ANSI/ISEA Z308.1-2009 standards.
Catalog No. 50-008-27883 Supplier Enterprise Technology Solutions Supplier No. FAOAN205
All-purpose trauma or combine dressing pad is ideal to stop bleeding associated with deep lacerations, abrasions, burns, penetration wounds and fractures. Sterile pad is individually wrapped to prevent contamination. Color-coded, unitized refill is designed to fit neatly into unitized first aid stations or kits. It also meets ANSI/ISEA Z308.1-2009 standards.