Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MSC Medique Wound Care Ointment, Triple Antibiotic, 1/57 oz, 25 Pack

Catalog No. 501909436
Encompass

This wound care ointment is a triple antibiotic formulation, supplied in a box with a container size of 1/57 oz. It is designed for general wound care applications.

  • Form: Ointment
  • Container type: Box
  • Container size: 1/57 oz
  • Contains triple antibiotic ointment
  • Not unitized kit packaging
  • Country of origin: United States

Catalog No. 50-190-9436 Supplier MSC Supplier No. 45907912
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This wound care ointment is a triple antibiotic formulation, supplied in a box with a container size of 1/57 oz. It is designed for general wound care applications.

  • Form: Ointment
  • Container type: Box
  • Container size: 1/57 oz
  • Contains triple antibiotic ointment
  • Not unitized kit packaging
  • Country of origin: United States

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.