Learn More
MSC Namco Floor Sealer and Cleaner, Multi-Surface, High-Gloss Finish, 1 gal
Catalog No. 502819428
Encompass
Namco First Kote Floor Sealer and Multi-Purpose Cleaner is a versatile solution designed for treating and maintaining various hard surfaces. It functions as a long-lasting sealer, providing a high-gloss finish, and can also be used as an effective multi-purpose cleaner.
  • Must for porous surfaces, terrazzo tile, paneling, hardwood floors, and fixtures
  • Long-lasting sealer formula for a high-gloss finish
  • Use for the first coat before waxing
  • Can also be used as a multi-purpose cleaner
Catalog No. 50-281-9428 Supplier MSC Supplier No. 20691721
Namco First Kote Floor Sealer and Multi-Purpose Cleaner is a versatile solution designed for treating and maintaining various hard surfaces. It functions as a long-lasting sealer, providing a high-gloss finish, and can also be used as an effective multi-purpose cleaner.
  • Must for porous surfaces, terrazzo tile, paneling, hardwood floors, and fixtures
  • Long-lasting sealer formula for a high-gloss finish
  • Use for the first coat before waxing
  • Can also be used as a multi-purpose cleaner