Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hughes Modesty Curtains

Catalog No. 18999466B Shop All Justrite Safety Group Products
Change view
Click to view available options
Mounting:
Pipe
Wall
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Mounting
18-999-466B Wall
18-999-466A Pipe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-999-466B Supplier Justrite Manufacturing Co Supplier No. CURTAINWM
  • $802.00
    $623.65 / Each of 1

    Save $178.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Provide privacy during emergencies

  • For use with installed safety shower system
  • Two styles available, both designed to work with any commercially available safety shower
  • Pipe-mount for freestanding, plumbed-in safety showers; measure 52 in. dia.
  • Wall-mount for freestanding systems near a wall or wall-mounted plumbed system
  • Made of 20 oz. polyester fabric with nickel-cadmium plated rings
  • Epoxy-sealed steel supports exhibit exceptional resistance to harsh environments

Specifications

Color Yellow
For Use With (Equipment) Installed Safety Shower System
Material Polyester, Nickel Cadmium, Epoxy Sealed Steel
Mounting Wall
Product Type Modesty Curtain
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.