Learn More
NOWDiagnostics, Inc. First To Know Rapid Syphilis Test
Catalog No. 21377374
Change view
Click to view available options
Quantity:
1 Test
25 Test
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-377-374 1 Test
21-377-375 25 Test
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 21-377-374 Supplier NOWDiagnostics, Inc. Supplier No. 600736
Only null left
Add to Cart
Add to Cart
First To Know Rapid Syphilis test is the first FDA-cleared point-of-care test for self-collection and over-the-counter detection of Syphilis. This CLIA-waived test offers a quick and accurate result, bringing you a convenient new syphilis testing option.
  • FDA cleared, over-the-counter, CLIA waived, 15 minute result
  • Able to be performed by any users, qualitative result (positive = test line)
  • Only cleared product that has fingerstick and blood drop directly on the device, no separation or sample prep needed - just Touch, Tap, and Test.
  • Intended for symptomatic or asymptomatic patients with no prior history of syphilis infectionThe First To
  • Know Rapid Syphilis test is the only FDA cleared POCT device for self collect and OTC
Specifications
CE Marker Yes
Clia Complexity Waived
Detectable Analytes Syphilis RPR Antigen
Type Syphilis Test
Disposable Single-use
Quantity 1 Test
Form Ready-to-Use (RTU)