Learn More
NOWDiagnostics, Inc. First To Know Rapid Syphilis Test
Description
- FDA cleared, over-the-counter, CLIA waived, 15 minute result
- Able to be performed by any users, qualitative result (positive = test line)
- Only cleared product that has fingerstick and blood drop directly on the device, no separation or sample prep needed - just Touch, Tap, and Test.
- Intended for symptomatic or asymptomatic patients with no prior history of syphilis infectionThe First To
- Know Rapid Syphilis test is the only FDA cleared POCT device for self collect and OTC
Specifications
Specifications
| CE Marker | CE-IVD |
| Clia Complexity | Waived |
| Detectable Analytes | Syphilis RPR Antigen |
| Type | Syphilis Test |
| Disposable | Single-use |
| Quantity | 25 Test |
| Form | Ready-to-Use (RTU) |