Learn More
Carbosynth LLC. H-Phe-Ile-Arg-Val-Val-Met-Tyr-Glu-Gly-Lys-Lys-OH, 1mg, MW: 1369.7, Molecular formula: C64H104N16O15S
SDP
Supplier:  Carbosynth LLC. TSP3873PI1MG
Encompass_Preferred
FIRVVMYEGKK, Application: CD47 binding peptide derived from thrombospondin 1; inhibitor of angiogenic responses, Salt Form:, Trifluoroacetate Form, Storage Temperature: -20 °C, References: S. Kaur et al., J. Biol. Chem., 285: 38923 (2010).
Catalog No. 50-168-7205