Learn More
AdipoGen Fis1 pAb
SDP
Supplier:  AdipoGen AG25B0007VC100
Encompass_Preferred
Polyclonal Antibody. Recognizes human, mouse and rat Fis1. Source Rabbit. Applications ICC, IP, WB. Liquid. In PBS containing 10% glycerol and 0.02% sodium azide. Fis1 promotes the fragmentation of the mitochondrial network and its perinuclear clustering. It can induce cytochrome c release from mitochondria to the cytosol, ultimately leading to apoptosis. It also mediates peroxisomal fission.
Catalog No. 50-256-3200