Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Animal Kingdom Poster
SCI_ED

Catalog No. S01954L Shop All Fischer Technical Company Products
Change view
Click to view available options
Format:
Poster
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S01954L Poster
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01954L Supplier Fischer Technical Company Supplier No. A111L

missing translation for 'validMainframeMsg'

Presenting the new cladistics classification system with 6 newly established phyla/clades being added, as well as 11 containing only extinct species.

  • Essential reference chart for teachers, students and professionals dealing with both living and prehistoric species
  • Three poster Set: Animal Development, Milestones of Evolution and Our New Animal Kingdom 2 are designed to be used both individually and together

Specifications

Format Poster
Product Type Laminated Animal Kingdom Poster
For Use With (Application) Essential reference chart for teachers, students and professionals
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.