Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Beaufort Wind Force Scale Poster
SCI_ED

Catalog No. S01959L Shop All Fischer Technical Company Products
Change view
Click to view available options
Flat; Legend: Beaufort Wind Force Scale
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S01959L Flat; Legend: Beaufort Wind Force Scale
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01959L Supplier Fischer Technical Company Supplier No. E111L

missing translation for 'validMainframeMsg'

This poster cites the official description of each wind force, but goes beyond that to show how it impacts both land and sea.

  • Illustrations show increasingly powerful winds and waves systemically destroying a summer beach cottage and tearing off large sections of the cliff, severely eroding the shoreline
  • Used to determine wind speed based solely on observation
  • Essential reference chart for meteorologists, seamen or anyone else who wants or needs to know how to identify weather conditions
  • Size: 24 x 36 in.

Specifications

Format Poster
Product Type Beaufort Wind Force Scale
Width (English) 36 in.
Height (English) 24 in.
For Use With (Application) Used to determine wind speed based solely on observation
Subject Physics
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.