Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Butterflies of the World Poster
SCI_ED

Catalog No. S01962L Shop All Fischer Technical Company Products
Change view
Click to view available options
Format:
Poster
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S01962L Poster
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01962L Supplier Fischer Technical Company Supplier No. A221L

missing translation for 'validMainframeMsg'

The Lepidoptera order/clade contains the butterflies and moth.

  • Over 100 of the most colorful species are shown
  • The captions cite common name, biological name and where they are found
  • These beautiful insects are grouped by continent or other geographic area.
  • Unique globe and arrow icons identify the location

Specifications

Format Poster
Product Type Laminated Butterflies of the World Poster
For Use With (Application) The Lepidoptera order/clade contains the butterflies and moth
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.