Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Insect Identification Poster
SCI_ED

Catalog No. S01963L Shop All Fischer Technical Company Products
Change view
Click to view available options
Format:
Poster
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Format
S01963L Poster
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S01963L Supplier Fischer Technical Company Supplier No. A222L

missing translation for 'validMainframeMsg'

Extremely popular with collectors, kids and educators. It is widely used in schools and by collectors.

  • Presents all the insect orders through a brief description accompanied by detailed pictures of one or more species
  • Glossary provides information on each body part
  • Includes a section that introduces Arthropods
  • A section on anatomy identifies important physical characteristics

Specifications

Format Poster
Product Type Insect Identification Poster
Includes A section that introduces Arthropods
For Use With (Application) Widely used in schools and by collectors
Subject Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.