Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Introduction to Gem Stones Poster
SCI_ED

Catalog No. S04768 Shop All Fischer Technical Company Products
Change view
Click to view available options
Introduction to Gem Stones poster; 24 x 36 in. (61 x 91.4cm)
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
S04768 Introduction to Gem Stones poster; 24 x 36 in. (61 x 91.4cm)
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S04768 Supplier Fischer Technical Company Supplier No. E107L

missing translation for 'validMainframeMsg'

Physical and chemical properties displayed in easy-to-see rows

  • Provides explanation of cut, clarity, color and caret
  • Grouped together by class
  • Properties such as asterism, opalescence, hardness and pattern shown
  • Birthstones, jewelry and non-jewelry uses listed
  • Laminated
  • Size: 24 x 36 in. (61 x 91.4cm)

Specifications

Product Type Introduction to Gem Stones Poster
Includes Physical and chemical properties, jewelry and non-jewelry uses, information about birthstones
For Use With (Application) Provides explanation of cut, clarity, color and caret, properties such as asterism, opalescence, hardness and pattern shown
Subject Science
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.