Learn More
Indofine Chemical Fisetin, 98%, 500 Mg
SDP
Catalog No. 50908302
Encompass_Preferred
Molecular Formula: C15H10O6, Purity: 98%, Molecular Weight: 286.25, Appearance: Light yellow powder, Solubility: Soluble in DMSO, Storage conditions: Store in a cool, dry place, Also known as 3,7,3',4'-Tetrahydroxyflavone
Catalog No. 50-908-302 Supplier Indofine Chemical Supplier No. F002 500 MG
Molecular Formula: C15H10O6, Purity: 98%, Molecular Weight: 286.25, Appearance: Light yellow powder, Solubility: Soluble in DMSO, Storage conditions: Store in a cool, dry place, Also known as 3,7,3',4'-Tetrahydroxyflavone