Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisetin (Tech.), 70%, Thermo Scientific™

Catalog No. AC119155000
Change view
Click to view available options
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
AC119155000 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC119155000 Supplier Thermo Scientific Chemicals Supplier No. 119155000

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 528-48-3
Infrared Spectrum Authentic
Assay Percent Range 70%
Molecular Formula C15H10O6
Quantity 500mg
Beilstein 18, 221
Merck Index 15, 4119
Synonym fisetin, viset, 5-desoxyquercetin, fustel, 2-3,4-dihydroxyphenyl-3,7-dihydroxy-4h-chromen-4-one, fisetholz, superfustel, cotinin, fietin, fustet
InChI Key XHEFDIBZLJXQHF-UHFFFAOYSA-N
SMILES C1=CC(=C(C=C1C2=C(C(=O)C3=C(O2)C=C(C=C3)O)O)O)O
IUPAC Name 2-(3,4-dihydroxyphenyl)-3,7-dihydroxychromen-4-one
Molecular Weight (g/mol) 286.239
PubChem CID 5281614
ChEBI CHEBI:42567
Formula Weight 286.23
Percent Purity ≥65%
Grade Technical
Chemical Name or Material Fisetin, tech., 70%
Show More Show Less
EINECSNumber 208-434-4
RTECSNumber LK9250000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.