Learn More
American Research Products Inc Fish Acetylcholinesterase ACh
Catalog No. 5000328572
Encompass
Fish Acetylcholinesterase (AChE) ELISA Kit from CUSABIO Detection Range 2 5 ng/mL-40 ng/mL Sensitivity 2 5 ng/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28572 Supplier American Research Products Inc Supplier No. CSBE17001FH
Fish Acetylcholinesterase (AChE) ELISA Kit from CUSABIO Detection Range 2 5 ng/mL-40 ng/mL Sensitivity 2 5 ng/mL Sample serum plasma tissue homogenates Method Competitive