Learn More
American Research Products Inc Fish adrenocorticotropic hormo
Catalog No. 5000328546
Encompass
Fish adrenocorticotropic hormone (ACTH) ELISA kit from CUSABIO Detection Range 75 pg/mL-1200 pg/mL Sensitivity 75 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28546 Supplier American Research Products Inc Supplier No. CSBE15926FH
Fish adrenocorticotropic hormone (ACTH) ELISA kit from CUSABIO Detection Range 75 pg/mL-1200 pg/mL Sensitivity 75 pg/mL Sample serum plasma tissue homogenates Method Competitive