Learn More
Cusabio Technology LLC Fish Catalase CAT ELISA kit
Catalog No. 5000051991
Encompass_Preferred
The Fish Catalase (CAT) ELISA Kit is designed for the quantitative detection of catalase (CAT) in fish serum and plasma samples using a competitive ELISA method This kit detects CAT within a broad dynamic range of 125 mU/mL to 2000 mU/mL with a sensitivity of 62 5 mU/mL ensuring precise measurement of enzyme activity The assay requires a short incubation period of 1 to 5 hours and utilizes a minimal sample volume of 50-100 L per test Optical density readings are performed at 450 nm providing reliable and reproducible results for research applications Ideal for studying oxidative stress biomarkers this kit offers a robust and efficient tool for quantifying CAT levels in aquatic species
Catalog No. 50-000-51991 Supplier Cusabio Technology LLC Supplier No. CSBE15928FH
The Fish Catalase (CAT) ELISA Kit is designed for the quantitative detection of catalase (CAT) in fish serum and plasma samples using a competitive ELISA method This kit detects CAT within a broad dynamic range of 125 mU/mL to 2000 mU/mL with a sensitivity of 62 5 mU/mL ensuring precise measurement of enzyme activity The assay requires a short incubation period of 1 to 5 hours and utilizes a minimal sample volume of 50-100 L per test Optical density readings are performed at 450 nm providing reliable and reproducible results for research applications Ideal for studying oxidative stress biomarkers this kit offers a robust and efficient tool for quantifying CAT levels in aquatic species