Learn More
Medchemexpress LLC Fish gelatin | 9000-70-8 | 250mg
SDP
Catalog No. 5000266667
Encompass_Preferred
Fish Gelatin suitable for cell culture is a gelatin from the skin of cold-water fish with low gelling and melting points which can be used for cell culture[1]
Catalog No. 50-002-66667 Supplier Medchemexpress LLC Supplier No. HYY1365C250MG
Fish Gelatin suitable for cell culture is a gelatin from the skin of cold-water fish with low gelling and melting points which can be used for cell culture[1]