Learn More
American Research Products Inc Fish Glutathione PeroXidase G
Catalog No. 5000328610
Encompass
Fish Glutathione PeroXidase (GSH-PX) ELISA kit from CUSABIO Detection Range 375 U/mL-6000 U/mL Sensitivity 187 5 U/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28610 Supplier American Research Products Inc Supplier No. CSBE15930FH
Fish Glutathione PeroXidase (GSH-PX) ELISA kit from CUSABIO Detection Range 375 U/mL-6000 U/mL Sensitivity 187 5 U/mL Sample serum plasma tissue homogenates Method Competitive