Learn More
American Research Products Inc Fish Heat Shock Protein 70 HS
Catalog No. 5000328620
Encompass
Fish Heat Shock Protein 70 (HSP-70) ELISA Kit from CUSABIO Detection Range 18 75 pg/mL-300 pg/mL Sensitivity 18 75 pg/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28620 Supplier American Research Products Inc Supplier No. CSBE16327FH
Fish Heat Shock Protein 70 (HSP-70) ELISA Kit from CUSABIO Detection Range 18 75 pg/mL-300 pg/mL Sensitivity 18 75 pg/mL Sample serum plasma tissue homogenates Method Competitive