Learn More
American Research Products Inc Fish hepatocyte growth factor
Catalog No. 5000328613
Encompass
Fish hepatocyte growth factor (HGF) ELISA Kit from CUSABIO Detection Range 230 pg/mL-8000 pg/mL Sensitivity 145 pg/mL Sample serum plasma Method Competitive
Catalog No. 50-003-28613 Supplier American Research Products Inc Supplier No. CSBE16527FH
Fish hepatocyte growth factor (HGF) ELISA Kit from CUSABIO Detection Range 230 pg/mL-8000 pg/mL Sensitivity 145 pg/mL Sample serum plasma Method Competitive