Learn More
American Research Products Inc Fish Insulin like growth facto
Catalog No. 5000327705
Encompass
Fish Insulin-like growth factor II (IGF2) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-1000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma Method Competitive
Catalog No. 50-003-27705 Supplier American Research Products Inc Supplier No. CSBEL011088FI
Fish Insulin-like growth factor II (IGF2) ELISA kit from CUSABIO Detection Range 62 5 pg/mL-1000 pg/mL Sensitivity 31 25 pg/mL Sample serum plasma Method Competitive