Learn More
American Research Products Inc Fish Interleukin 1b IL 1b E
Supplier:  American Research Products Inc CSBE13259FH
Encompass
Fish Interleukin 1b (IL- 1b) ELISA Kit from CUSABIO Detection Range 3 12 pg/mL-200 pg/mL Sensitivity 0 78 pg/mL Sample serum plasma tissue homogenates Method Sandwich
Catalog No. 50-003-26625