Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fish Oil, From Menhaden, Spectrum™ Chemical
SDP

Catalog No. 18602734 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
5 gal.
Packaging:
Amber Glass Bottle
Poly Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-602-734 500 mL Amber Glass Bottle
18-602-735 5 gal. Poly Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 18-602-734 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. F1192500ML
Add to Cart
Add to Cart

F1192, 8002-50-4

Spectrum™ Chemical Fish Oil, From Menhaden is an oil obtained from the small North Atlantic fish, Brevoortia tyrannus. Fish oil derived from Menhaden boast low levels of mercury while still being high in omega-3 fatty acid. It is used in cosmetic and personal care products as a skin-conditioning agent, emollient and solvent. It appears as a liquid that is insoluble in cold and hot water.

Chemical Identifiers

CAS 8002-50-4

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 500 mL
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.