Learn More
American Research Products Inc Fish thyroid stimulating hormo
Catalog No. 5000328311
Encompass
Fish thyroid stimulating hormone (TSH) ELISA Kit from CUSABIO Detection Range 2 5 IU/mL-40 IU/mL Sensitivity 2 5 IU/mL Sample serum plasma tissue homogenates Method Competitive
Catalog No. 50-003-28311 Supplier American Research Products Inc Supplier No. CSBEQ027261FI
Fish thyroid stimulating hormone (TSH) ELISA Kit from CUSABIO Detection Range 2 5 IU/mL-40 IU/mL Sensitivity 2 5 IU/mL Sample serum plasma tissue homogenates Method Competitive