Learn More
American Research Products Inc Fish transforming growth facto
Catalog No. 5000328246
Encompass
Fish transforming growth factor beta 1 (TGFB1) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 3 12 ng/mL Sample serum plasma Method Sandwich
Catalog No. 50-003-28246 Supplier American Research Products Inc Supplier No. CSBEL023446FI
Fish transforming growth factor beta 1 (TGFB1) ELISA kit from CUSABIO Detection Range 6 25 ng/mL-400 ng/mL Sensitivity 3 12 ng/mL Sample serum plasma Method Sandwich